Assay Details
Target Gene Details
Entrez Gene ID: | 91283 |
Gene Name: | Myb/SANT DNA binding domain containing 3 |
Gene Aliases: |
C9orf30, L8 |
Location: |
Chr.9:100427213-100451734 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSANTD3 | NM_001198805.1 | NP_001185734.1 | ||
| NM_001198806.1 | NP_001185735.1 | |||
| NM_001198807.1 | NP_001185736.1 | |||
| NM_080655.2 | NP_542386.1 | |||
| AK314924.1 | ||||
| BC008993.1 | AAH08993.1 | |||
| BI596498.1 | ||||
| BI600290.1 | ||||
| BM914752.1 | ||||
| CN366267.1 | ||||
| DA424604.1 | ||||
| DR159535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825012 | Chr.9:100389923 - 100493999 on Build GRCh38 | Gain |
|
| nsv614955 | Chr.9:100419314 - 100498945 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map