Assay Details
Target Gene Details
Entrez Gene ID: | 6136 |
Gene Name: | ribosomal protein L12 |
Gene Aliases: |
L12 |
Location: |
Chr.9:127447674-127451432 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL12 | NM_000976.3 | NP_000967.1 | ||
| AF037643.1 | ||||
| AK057602.1 | 1 | 1202 | ||
| CB155565.1 | ||||
| L06505.1 | AAA36157.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75945869] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map