Assay Details
Target Gene Details
| Entrez Gene ID: | 105376204 | 
| Gene Name: | uncharacterized LOC105376204 | 
| Gene Aliases: | - | 
| Location: | Chr.9:106852964-106863015 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| LOC105376204 | XM_017015409.1 | 1 | 253 | XP_016870898.1 | 
Target Gene Details
| Entrez Gene ID: | 58499 | 
| Gene Name: | zinc finger protein 462 | 
| Gene Aliases: | Zfp462 | 
| Location: | Chr.9:106860154-107012258 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| ZNF462 | XM_006717212.3 | XP_006717275.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Intronic  Non-exonic  non-DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top