Assay Details
Target Gene Details
Entrez Gene ID: | 4712 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B6 |
Gene Aliases: |
B17, CI |
Location: |
Chr.9:32553526-32573184 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFB6 | NM_001199987.1 | NP_001186916.1 | ||
| NM_002493.4 | NP_002484.1 | |||
| NM_182739.2 | NP_877416.1 | |||
| AF035840.1 | AAC68838.1 | |||
| AF067167.1 | AAD32451.1 | |||
| AK289702.1 | ||||
| AW160372.1 | ||||
| BC009801.1 | AAH09801.1 | |||
| BC029247.1 | AAH29247.1 | |||
| BI669356.1 | ||||
| BP208846.1 | ||||
| BU952486.1 | ||||
| CR456920.1 | CAG33201.1 |
Target Gene Details
Entrez Gene ID: | 100129250 |
Gene Name: | TOPORS antisense RNA 1 |
Gene Aliases: |
C9orf133 |
Location: |
Chr.9:32551144-32566860 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOPORS-AS1 | NR_102376.1 | |||
| NR_102377.1 | ||||
| BI916035.1 | ||||
| DA702677.1 | ||||
| DA753662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620302 | Chr.9:32556081 - 32559949 on Build GRCh38 | Gain |
|
| nsv831540 | Chr.9:32509387 - 32651746 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map