Assay Details
Target Gene Details
Entrez Gene ID: | 5253 |
Gene Name: | PHD finger protein 2 |
Gene Aliases: |
CENP-35, GRC5, JHDM1E |
Location: |
Chr.9:93576627-93679587 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHF2 | NM_005392.3 | NP_005383.3 | ||
XM_005252051.2 | XP_005252108.1 | |||
XM_006717143.2 | XP_006717206.1 | |||
AF043725.1 | AAD21791.1 | |||
AL834263.1 | CAD38938.1 | |||
BC064613.1 | AAH64613.1 | |||
CB990295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509311 | Chr.9:93580555 - 93696735 on Build GRCh38 | Insertion |
![]() ![]() ![]() |
nsv824984 | Chr.9:93548651 - 93612740 on Build GRCh38 | Gain |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
