Assay Details
Target Gene Details
Entrez Gene ID: | 79956 |
Gene Name: | endoplasmic reticulum metallopeptidase 1 |
Gene Aliases: |
FXNA, KIAA1815, bA207C16.3 |
Location: |
Chr.9:5764061-5867091 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERMP1 | XM_017015139.1 | XP_016870628.1 |
Target Gene Details
Entrez Gene ID: | 158358 |
Gene Name: | KIAA2026 |
Gene Aliases: |
- |
Location: |
Chr.9:5860254-6008479 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA2026 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv613203 | Chr.9:5841438 - 5877948 on Build GRCh38 | Loss |
|
| nsv1029665 | Chr.9:5687332 - 6775107 on Build GRCh38 | Gain |
|
| nsv6463 | Chr.9:5836620 - 5870203 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map