Assay Details
Target Gene Details
Entrez Gene ID: | 105376204 |
Gene Name: | uncharacterized LOC105376204 |
Gene Aliases: |
- |
Location: |
Chr.9:106852964-106863015 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105376204 | XM_017015409.1 | 1 | 5685 | XP_016870898.1 |
