Assay Details
Target Gene Details
Entrez Gene ID: | 56262 |
Gene Name: | leucine rich repeat containing 8 family member A |
Gene Aliases: |
AGM5, LRRC8, SWELL1 |
Location: |
Chr.9:128882112-128918042 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC8A | NM_001127244.1 | NP_001120716.1 | ||
| NM_001127245.1 | NP_001120717.1 | |||
| NM_019594.3 | NP_062540.2 | |||
| XM_005252095.2 | XP_005252152.1 | |||
| XM_005252096.3 | 1 | 1005 | XP_005252153.1 | |
| XM_006717186.2 | XP_006717249.1 | |||
| XM_011518863.1 | XP_011517165.1 | |||
| XM_011518864.2 | XP_011517166.1 | |||
| AB037858.1 | ||||
| AK289842.1 | ||||
| BC051322.1 | AAH51322.1 | |||
| BX376144.2 | ||||
| DA243590.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621819 | Chr.9:128877077 - 128887549 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map