Assay Details
Target Gene Details
Entrez Gene ID: | 51531 |
Gene Name: | tRNA methyltransferase O |
Gene Aliases: |
C9orf156, HSPC219, NAP1 |
Location: |
Chr.9:97876321-97922570 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMO | XM_005252047.3 | XP_005252104.1 | ||
| XM_005252048.4 | 6 | 3941 | XP_005252105.1 | |
| XM_005252049.4 | 4 | 3663 | XP_005252106.1 | |
| XM_005252050.4 | 5 | 3767 | XP_005252107.1 | |
| XM_011518776.2 | XP_011517078.1 |
More Information
Set Membership: |
|

Genomic Map