Assay Details
Target Gene Details
Entrez Gene ID: | 114789 |
Gene Name: | solute carrier family 25 member 25 |
Gene Aliases: |
MCSC, PCSCL, SCAMC-2 |
Location: |
Chr.9:128068200-128109258 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A25 | NM_001006641.3 | NP_001006642.1 | ||
XM_005251688.1 | XP_005251745.1 | |||
AJ619990.1 | CAF04496.1 | |||
AK290991.1 | ||||
AL559911.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1054891 | Chr.9:127939274 - 128102397 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv471324 | Chr.9:128039877 - 128130666 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
