Assay Details
Target Gene Details
Entrez Gene ID: | 10868 |
Gene Name: | ubiquitin specific peptidase 20 |
Gene Aliases: |
LSFR3A, VDU2, hVDU2 |
Location: |
Chr.9:129835417-129881838 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP20 | NM_001008563.4 | NP_001008563.2 | ||
| NM_001110303.3 | NP_001103773.2 | |||
| NM_006676.7 | NP_006667.3 | |||
| XM_005251665.3 | XP_005251722.1 | |||
| XM_011518161.2 | XP_011516463.1 | |||
| XM_011518162.2 | 1 | 449 | XP_011516464.1 | |
| AY074877.1 | AAL79676.1 | |||
| BC039593.1 | AAH39593.1 | |||
| BF727180.1 | ||||
| BP246486.1 | ||||
| DA791069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046477 | Chr.9:129608081 - 129997996 on Build GRCh38 | Gain |
|
| esv3621839 | Chr.9:129701704 - 129885823 on Build GRCh38 | Loss |
|
| nsv1161909 | Chr.9:129834501 - 129870679 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map