Assay Details
Target Gene Details
Entrez Gene ID: | 48 |
Gene Name: | aconitase 1 |
Gene Aliases: |
ACONS, HEL60, IREB1, IREBP, IREBP1, IRP1 |
Location: |
Chr.9:32384603-32450836 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACO1 | NM_001278352.1 | NP_001265281.1 | ||
| NM_002197.2 | NP_002188.1 | |||
| XM_005251476.1 | XP_005251533.1 | |||
| AB209480.1 | BAD92717.1 | |||
| AF261088.1 | AAF99681.1 | |||
| AK057904.1 | ||||
| AK308752.1 | ||||
| BP287929.1 | ||||
| DA103947.1 | ||||
| DA409113.1 | ||||
| DA780353.1 | ||||
| Z11559.1 | CAA77651.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020675 | Chr.9:32323273 - 32391249 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map