Assay Details
Target Gene Details
Entrez Gene ID: | 9414 |
Gene Name: | tight junction protein 2 |
Gene Aliases: |
C9DUPq21.11, DFNA51, DUP9q21.11, PFIC4, X104, ZO2 |
Location: |
Chr.9:69099907-69255208 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TJP2 | XM_017015326.1 | XP_016870815.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508550 | Chr.9:69064250 - 69194517 on Build GRCh38 | Deletion |
|
| nsv6550 | Chr.9:69099228 - 69145132 on Build GRCh38 | Deletion |
|
| dgv7667n100 | Chr.9:69071560 - 69103531 on Build GRCh38 | Gain |
|
| esv3891679 | Chr.9:68939243 - 69246082 on Build GRCh38 | Gain |
|
| esv3576657 | Chr.9:68955164 - 69227475 on Build GRCh38 | Gain |
|
| dgv7666n100 | Chr.9:68896889 - 69228107 on Build GRCh38 | Gain |
|
| esv3891678 | Chr.9:68918451 - 69144626 on Build GRCh38 | Gain |
|
| nsv614548 | Chr.9:68856996 - 69320661 on Build GRCh38 | Gain |
|
| esv2761531 | Chr.9:69069453 - 69103543 on Build GRCh38 | Gain |
|
| dgv7665n100 | Chr.9:68783797 - 69356369 on Build GRCh38 | Gain |
|
| nsv466397 | Chr.9:68959841 - 69228107 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116717130] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map