Assay Details
Target Gene Details
Entrez Gene ID: | 158358 |
Gene Name: | KIAA2026 |
Gene Aliases: |
- |
Location: |
Chr.9:5860254-6008479 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA2026 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1029665 | Chr.9:5687332 - 6775107 on Build GRCh38 | Gain |
|
| nsv1033546 | Chr.9:5875201 - 5900427 on Build GRCh38 | Loss |
|

Genomic Map