Assay Details
Target Gene Details
Entrez Gene ID: | 65083 |
Gene Name: | nucleolar protein 6 |
Gene Aliases: |
NRAP, UTP22, bA311H10.1 |
Location: |
Chr.9:33460811-33473943 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOL6 | XM_017015044.1 | 26 | 4937 | XP_016870533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516500 | Chr.9:33434933 - 33466861 on Build GRCh38 | Loss |
|
| nsv1024207 | Chr.9:33296370 - 33574894 on Build GRCh38 | Gain |
|
| esv3576648 | Chr.9:33283584 - 33531049 on Build GRCh38 | Gain |
|
| dgv7517n100 | Chr.9:33380150 - 33468684 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map