Assay Details
Target Gene Details
Entrez Gene ID: | 4957 |
Gene Name: | outer dense fiber of sperm tails 2 |
Gene Aliases: |
CT134, ODF2/1, ODF2/2, ODF84 |
Location: |
Chr.9:128455155-128501292 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ODF2 | NM_153436.1 | NP_702914.1 | ||
| XM_011518729.2 | XP_011517031.1 | |||
| XM_011518732.2 | XP_011517034.1 | |||
| XM_017014774.1 | XP_016870263.1 | |||
| AK301842.1 | ||||
| DC397869.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78063361] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map