Assay Details
Target Gene Details
Entrez Gene ID: | 5288 |
Gene Name: | phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma |
Gene Aliases: |
PI3K-C2-gamma, PI3K-C2GAMMA |
Location: |
Chr.12:18242946-18716861 on Build GRCh38 |
Assay Gene Location: | Within Intron 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3C2G | XM_011520696.2 | XP_011518998.1 | ||
| XM_017019470.1 | XP_016874959.1 | |||
| XM_017019471.1 | XP_016874960.1 | |||
| XM_017019472.1 | XP_016874961.1 | |||
| XM_017019473.1 | XP_016874962.1 | |||
| XM_017019474.1 | XP_016874963.1 | |||
| XM_017019476.1 | XP_016874965.1 | |||
| XM_017019477.1 | XP_016874966.1 | |||
| XM_017019479.1 | XP_016874968.1 |
Target Gene Details
Entrez Gene ID: | 89869 |
Gene Name: | phospholipase C zeta 1 |
Gene Aliases: |
NYD-SP27, PLC-zeta-1, PLCzeta |
Location: |
Chr.12:18645529-18738059 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCZ1 | NM_033123.3 | NP_149114.2 | ||
| NR_073075.1 | ||||
| XM_005253519.2 | XP_005253576.1 | |||
| XM_011520883.1 | XP_011519185.1 | |||
| XM_017020176.1 | XP_016875665.1 | |||
| XM_017020177.1 | XP_016875666.1 | |||
| XM_017020178.1 | XP_016875667.1 | |||
| XM_017020179.1 | XP_016875668.1 | |||
| XM_017020180.1 | XP_016875669.1 | |||
| XM_017020181.1 | XP_016875670.1 | |||
| XM_017020182.1 | XP_016875671.1 | |||
| XM_017020183.1 | XP_016875672.1 | |||
| XM_017020184.1 | XP_016875673.1 | |||
| XM_017020185.1 | XP_016875674.1 | |||
| XM_017020186.1 | XP_016875675.1 | |||
| AF532185.1 | AAN71895.1 | |||
| AK097555.1 | BAC05099.1 | |||
| AK097725.1 | BAC05152.1 | |||
| AK292279.1 | ||||
| AK314516.1 | ||||
| AY035866.1 | AAK61372.1 | |||
| BC125067.1 | ||||
| HY197954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557672 | Chr.12:18654397 - 18698600 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map