Assay Details
Target Gene Details
Entrez Gene ID: | 6334 |
Gene Name: | sodium voltage-gated channel alpha subunit 8 |
Gene Aliases: |
CERIII, CIAT, EIEE13, MED, NaCh6, Nav1.6, PN4 |
Location: |
Chr.12:51589959-51812865 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN8A | NM_001177984.2 | NP_001171455.1 | ||
| NM_014191.3 | NP_055006.1 | |||
| XM_006719556.3 | XP_006719619.1 | |||
| XM_011538650.2 | XP_011536952.1 | |||
| XM_011538651.2 | XP_011536953.1 | |||
| XM_017019794.1 | XP_016875283.1 | |||
| XM_017019795.1 | XP_016875284.1 | |||
| XM_017019796.1 | XP_016875285.1 | |||
| AB027567.1 | BAA78033.1 | |||
| AK309213.1 | ||||
| AY510076.1 | AAS89685.1 | |||
| AY510083.1 | AAS89692.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs112148094] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map