Assay Details
Target Gene Details
Entrez Gene ID: | 55089 |
Gene Name: | solute carrier family 38 member 4 |
Gene Aliases: |
ATA3, NAT3, PAAT, SNAT4 |
Location: |
Chr.12:46764761-46832422 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC38A4 | NM_001143824.1 | NP_001137296.1 | ||
| NM_018018.4 | NP_060488.2 | |||
| XM_005268997.2 | XP_005269054.1 | |||
| AB055003.1 | BAB84090.1 | |||
| AF193836.1 | AAK71508.1 | |||
| AF305814.1 | AAK43528.1 | |||
| AK001053.1 | BAA91481.1 | |||
| AK291168.1 | ||||
| BC069819.1 | AAH69819.1 | |||
| BC101827.1 | AAI01828.1 | |||
| BC104913.1 | AAI04914.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826361 | Chr.12:46765762 - 46807111 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map