Assay Details
Target Gene Details
Entrez Gene ID: | 338811 |
Gene Name: | family with sequence similarity 19 member A2, C-C motif chemokine like |
Gene Aliases: |
TAFA-2, TAFA2 |
Location: |
Chr.12:61708248-62192839 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM19A2 | NM_178539.4 | NP_848634.1 | ||
| XM_006719370.2 | XP_006719433.1 | |||
| XM_011538272.1 | XP_011536574.1 | |||
| XM_011538274.1 | XP_011536576.1 | |||
| AK123580.1 | ||||
| AL834160.1 | CAD38865.1 | |||
| AY325115.1 | AAP92407.1 | |||
| BC028403.1 | AAH28403.1 | |||
| BC040286.1 | AAH40286.1 | |||
| CR749367.1 | CAH18220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581660 | Chr.12:61685385 - 61733005 on Build GRCh38 | Gain |
|
| nsv1042044 | Chr.12:60862234 - 61803104 on Build GRCh38 | Gain |
|
| nsv520898 | Chr.12:61684989 - 61721384 on Build GRCh38 | Gain |
|
| nsv1039741 | Chr.12:61653069 - 62097271 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79172187] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map