Assay Details
Target Gene Details
Entrez Gene ID: | 283420 |
Gene Name: | C-type lectin domain family 9 member A |
Gene Aliases: |
CD370, DNGR-1, DNGR1, UNQ9341 |
Location: |
Chr.12:10030677-10066030 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC9A | NM_207345.3 | NP_997228.1 | ||
| AY358265.1 | AAQ88632.1 | |||
| EU339276.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524870 | Chr.12:10057112 - 10067286 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map