Assay Details
Target Gene Details
Entrez Gene ID: | 56994 |
Gene Name: | choline phosphotransferase 1 |
Gene Aliases: |
CPT, CPT1 |
Location: |
Chr.12:101697639-101752038 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHPT1 | NM_020244.2 | NP_064629.2 | ||
XM_011538574.1 | XP_011536876.1 | |||
AF047431.1 | AAD44019.1 | |||
AF195623.1 | AAF87947.1 | |||
AF195624.1 | AAF87948.1 | |||
AK075211.1 | ||||
AK093067.1 | ||||
AK226127.1 | ||||
BC020819.1 | AAH20819.1 | |||
BC050429.1 | AAH50429.1 | |||
CD300296.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832497 | Chr.12:101667198 - 101861862 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv859 | Chr.12:101696419 - 101720084 on Build GRCh38 | Deletion |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
