Assay Details
Target Gene Details
Entrez Gene ID: | 83892 |
Gene Name: | potassium channel tetramerization domain containing 10 |
Gene Aliases: |
BTBD28, MSTP028, ULRO61, hBACURD3 |
Location: |
Chr.12:109448655-109477544 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD10 | NM_001317395.1 | NP_001304324.1 | ||
| NM_001317399.1 | NP_001304328.1 | |||
| NM_031954.4 | NP_114160.1 | |||
| NR_133897.1 | ||||
| NR_133898.1 | ||||
| AB209358.1 | BAD92595.1 | |||
| AF113208.1 | AAG39279.1 | |||
| AK027543.1 | BAB55188.1 | |||
| AK056912.1 | ||||
| AK092170.1 | ||||
| AK123733.1 | ||||
| AK222548.1 | BAD96268.1 | |||
| AK309783.1 | ||||
| AY205299.1 | AAO47716.1 | |||
| AY597809.1 | AAT09002.1 | |||
| BC040062.1 | AAH40062.1 | |||
| BX333975.2 | ||||
| CX785497.1 |
More Information
Set Membership: |
|

Genomic Map