Assay Details
Target Gene Details
| Entrez Gene ID: | 775 | 
| Gene Name: | calcium voltage-gated channel subunit alpha1 C | 
| Gene Aliases: | CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS | 
| Location: | Chr.12:1969677-2697949 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 50 | 
Target Gene Details
| Entrez Gene ID: | 100652846 | 
| Gene Name: | CACNA1C antisense RNA 1 | 
| Gene Aliases: | - | 
| Location: | Chr.12:2676001-2691157 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CACNA1C-AS1 | NR_045725.1 | |||
| AK093746.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1047874 | Chr.12:2577599 - 2807322 on Build GRCh38 | Gain |  CACNA1C-AS1  CACNA1C  FKBP4  CACNA1C-AS2  LOC283440 | 
| nsv832308 | Chr.12:2576562 - 2690062 on Build GRCh38 | Loss |  CACNA1C-AS1  CACNA1C  CACNA1C-AS2 | 
| dgv1344n100 | Chr.12:2673632 - 2740600 on Build GRCh38 | Gain |  CACNA1C-AS1  CACNA1C | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top