Assay Details
Target Gene Details
Entrez Gene ID: | 9815 |
Gene Name: | GIT ArfGAP 2 |
Gene Aliases: |
CAT-2, CAT2, PKL |
Location: |
Chr.12:109929792-109996389 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GIT2 | NM_001135213.1 | NP_001128685.1 | ||
| NM_001135214.1 | NP_001128686.1 | |||
| NM_014776.3 | NP_055591.2 | |||
| NM_057169.3 | NP_476510.1 | |||
| NM_057170.3 | NP_476511.1 | |||
| XM_005253997.3 | XP_005254054.1 | |||
| XM_005254000.3 | XP_005254057.1 | |||
| XM_006719707.3 | XP_006719770.1 | |||
| XM_006719708.3 | XP_006719771.1 | |||
| XM_006719709.3 | XP_006719772.1 | |||
| XM_006719712.3 | XP_006719775.1 | |||
| XM_017020258.1 | XP_016875747.1 | |||
| XM_017020259.1 | XP_016875748.1 | |||
| XM_017020260.1 | XP_016875749.1 | |||
| AF124491.1 | AAD28047.1 | |||
| AK303198.1 | ||||
| BC014223.2 | AAH14223.2 | |||
| CR749340.1 | CAH18193.1 | |||
| D63482.2 |
More Information
Set Membership: |
|

Genomic Map