Assay Details
Target Gene Details
Entrez Gene ID: | 2904 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 2B |
Gene Aliases: |
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3 |
Location: |
Chr.12:13537337-13982012 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIN2B | XM_005253351.3 | 4 | 13198 | XP_005253408.1 |
| XM_011520628.2 | 14 | 16264 | XP_011518930.1 | |
| XM_011520629.2 | 15 | 16071 | XP_011518931.1 | |
| XM_017019219.1 | 14 | 15819 | XP_016874708.1 |
More Information
Set Membership: |
|

Genomic Map