Assay Details
Target Gene Details
Entrez Gene ID: | 283461 |
Gene Name: | chromosome 12 open reading frame 40 |
Gene Aliases: |
HEL-206, HEL-S-94 |
Location: |
Chr.12:39626167-39908300 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C12orf40 | NR_135051.1 | |||
AK097445.1 | BAC05057.1 | |||
EU668334.1 | ||||
GQ472216.1 |
Target Gene Details
Entrez Gene ID: | 114134 |
Gene Name: | solute carrier family 2 member 13 |
Gene Aliases: |
HMIT |
Location: |
Chr.12:39755021-40106085 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC2A13 | NM_052885.3 | NP_443117.3 | ||
XM_011537847.2 | XP_011536149.1 | |||
XM_017018764.1 | XP_016874253.1 | |||
XM_017018765.1 | XP_016874254.1 | |||
XM_017018766.1 | XP_016874255.1 | |||
AJ315644.1 | CAC51116.1 | |||
BC117117.1 | ||||
BC117119.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428280 | Chr.12:39728911 - 39805819 on Build GRCh38 | Loss |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
