Assay Details
Target Gene Details
Entrez Gene ID: | 5781 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 11 |
Gene Aliases: |
BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2 |
Location: |
Chr.12:112418732-112509913 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN11 | NM_002834.3 | NP_002825.3 | ||
| NM_080601.1 | NP_542168.1 | |||
| XM_006719526.2 | XP_006719589.1 | |||
| XM_011538613.2 | XP_011536915.1 | |||
| XM_017019722.1 | XP_016875211.1 | |||
| AK289854.1 | ||||
| BC008692.1 | AAH08692.1 | |||
| BT007106.1 | AAP35770.1 | |||
| D13540.1 | BAA02740.2 | |||
| L03535.1 | AAA36611.1 | |||
| L07527.1 | AAA17022.1 | |||
| L08807.1 | ||||
| X70766.1 | CAA50045.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043127 | Chr.12:112419957 - 112462894 on Build GRCh38 | Gain |
|
| nsv1050197 | Chr.12:112456743 - 112470326 on Build GRCh38 | Gain |
|
| nsv889 | Chr.12:112445258 - 112490108 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map