Assay Details
Target Gene Details
Entrez Gene ID: | 55711 |
Gene Name: | fatty acyl-CoA reductase 2 |
Gene Aliases: |
HEL-S-81, MLSTD1, SDR10E2 |
Location: |
Chr.12:29149003-29335616 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAR2 | NM_001271783.1 | NP_001258712.1 | ||
| XM_017019624.1 | XP_016875113.1 | |||
| XM_017019625.1 | XP_016875114.1 | |||
| AK001324.1 | BAA91625.1 | |||
| AK314670.1 | ||||
| AL136843.1 | CAB66777.1 | |||
| DA068892.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951345 | Chr.12:29158268 - 29169367 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map