Assay Details
Target Gene Details
Entrez Gene ID: | 56938 |
Gene Name: | aryl hydrocarbon receptor nuclear translocator like 2 |
Gene Aliases: |
BMAL2, CLIF, MOP9, PASD9, bHLHe6 |
Location: |
Chr.12:27332854-27425813 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARNTL2 | NM_001248002.1 | NP_001234931.1 | ||
| NM_001248003.1 | NP_001234932.1 | |||
| NM_001248004.1 | NP_001234933.1 | |||
| NM_001248005.1 | NP_001234934.1 | |||
| NM_020183.4 | NP_064568.3 | |||
| XM_006719112.3 | XP_006719175.1 | |||
| XM_011520766.2 | XP_011519068.1 | |||
| XM_011520768.2 | XP_011519070.1 | |||
| XM_011520769.2 | XP_011519071.1 | |||
| XM_017019666.1 | XP_016875155.1 | |||
| XM_017019667.1 | XP_016875156.1 | |||
| AF246960.1 | AAL50339.1 | |||
| AF246961.1 | AAL50340.1 | |||
| AF246962.1 | AAL50341.1 | |||
| AF246963.1 | AAL50342.1 | |||
| AK296706.1 | ||||
| BC125061.1 | ||||
| BC125062.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1406n100 | Chr.12:27128236 - 27640822 on Build GRCh38 | Gain |
|
| dgv2417n54 | Chr.12:27068022 - 27638988 on Build GRCh38 | Gain |
|
| esv3628933 | Chr.12:27275994 - 27403117 on Build GRCh38 | Gain |
|
| nsv436825 | Chr.12:27135971 - 27642579 on Build GRCh38 | Insertion |
|
| esv2745664 | Chr.12:27017516 - 27820117 on Build GRCh38 | Deletion |
|
| nsv521872 | Chr.12:27114021 - 27635541 on Build GRCh38 | Gain |
|
| dgv271e199 | Chr.12:27089700 - 27631451 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map