Assay Details
Target Gene Details
Entrez Gene ID: | 283461 |
Gene Name: | chromosome 12 open reading frame 40 |
Gene Aliases: |
HEL-206, HEL-S-94 |
Location: |
Chr.12:39626167-39908300 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C12orf40 | NM_001031748.3 | NP_001026918.2 | ||
NM_001319247.1 | NP_001306176.1 | |||
NR_135051.1 | ||||
XM_005268806.3 | XP_005268863.1 | |||
XM_011538231.2 | XP_011536533.1 | |||
AK097445.1 | BAC05057.1 | |||
BC038754.1 | AAH38754.2 | |||
BC048120.1 | ||||
EU668334.1 | ||||
GQ472216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3629268 | Chr.12:39654619 - 39674444 on Build GRCh38 | Loss |
![]() |
esv3629269 | Chr.12:39667551 - 39701177 on Build GRCh38 | Loss |
![]() |
esv3629266 | Chr.12:39648921 - 39753228 on Build GRCh38 | Loss |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
