Assay Details
Target Gene Details
Entrez Gene ID: | 387893 |
Gene Name: | lysine methyltransferase 5A |
Gene Aliases: |
PR-Set7, SET07, SET8, SETD8 |
Location: |
Chr.12:123383778-123409357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KMT5A | NM_001324504.1 | NP_001311433.1 | ||
NM_001324505.1 | NP_001311434.1 | |||
NM_020382.4 | NP_065115.3 | |||
NR_136910.1 | ||||
XM_011538333.2 | XP_011536635.1 | |||
AK292645.1 | ||||
AY102937.1 | AAM47033.1 | |||
BC050346.1 | AAH50346.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv826534 | Chr.12:123375063 - 123391807 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
