Assay Details
Target Gene Details
Entrez Gene ID: | 7296 |
Gene Name: | thioredoxin reductase 1 |
Gene Aliases: |
GRIM-12, TR, TR1, TRXR1, TXNR |
Location: |
Chr.12:104215779-104350307 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXNRD1 | NM_001093771.2 | NP_001087240.1 | ||
| AK304241.1 | ||||
| AY057105.1 | AAL15432.1 | |||
| BG772375.1 | ||||
| DC418649.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630632 | Chr.12:104216440 - 104227182 on Build GRCh38 | Gain |
|
| esv3630630 | Chr.12:104203244 - 104250924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map