Assay Details
Target Gene Details
Entrez Gene ID: | 196463 |
Gene Name: | phospholipase B domain containing 2 |
Gene Aliases: |
P76 |
Location: |
Chr.12:113358566-113391582 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLBD2 | NM_001159727.1 | NP_001153199.1 | ||
| NM_173542.3 | NP_775813.2 | |||
| XM_011538023.2 | XP_011536325.1 | |||
| XM_017018977.1 | XP_016874466.1 | |||
| BC030618.2 | AAH30618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map