Assay Details
Target Gene Details
Entrez Gene ID: | 113675 |
Gene Name: | serine dehydratase like |
Gene Aliases: |
SDH 2, SDS-RS1, TDH, cSDH |
Location: |
Chr.12:113422265-113438277 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDSL | NM_001304993.1 | NP_001291922.1 | ||
| NM_138432.3 | NP_612441.1 | |||
| XM_005253831.4 | XP_005253888.1 | |||
| XM_011537846.2 | XP_011536148.1 | |||
| XM_017018763.1 | XP_016874252.1 | |||
| AF134473.1 | AAP97250.1 | |||
| BC009849.2 | AAH09849.1 | |||
| BC091479.1 | AAH91479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map