Assay Details
Target Gene Details
| Entrez Gene ID: | 55711 | 
| Gene Name: | fatty acyl-CoA reductase 2 | 
| Gene Aliases: | HEL-S-81, MLSTD1, SDR10E2 | 
| Location: | Chr.12:29149003-29335616 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| FAR2 | NM_001271783.1 | NP_001258712.1 | ||
| XM_017019624.1 | XP_016875113.1 | |||
| XM_017019625.1 | XP_016875114.1 | |||
| AK001324.1 | BAA91625.1 | |||
| AK314670.1 | ||||
| AL136843.1 | CAB66777.1 | |||
| DA068892.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv437739 | Chr.12:29176572 - 29185933 on Build GRCh38 | Loss |  FAR2 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top