Assay Details
Target Gene Details
Entrez Gene ID: | 5801 |
Gene Name: | protein tyrosine phosphatase, receptor type R |
Gene Aliases: |
EC-PTP, PCPTP1, PTP-SL, PTPBR7, PTPRQ |
Location: |
Chr.12:70638073-70920804 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRR | NM_001207015.1 | NP_001193944.1 | ||
| NM_001207016.1 | NP_001193945.1 | |||
| NM_002849.3 | NP_002840.2 | |||
| NM_130846.2 | NP_570897.2 | |||
| NR_073474.1 | ||||
| XM_011538615.2 | XP_011536917.1 | |||
| AK091647.1 | ||||
| AK295951.1 | ||||
| AK304457.1 | ||||
| AK312376.1 | ||||
| BC110900.1 | AAI10901.1 | |||
| BC119777.1 | ||||
| BC122866.1 | ||||
| BX571751.1 | CAE11876.1 | |||
| CR749836.1 | CAH18692.1 | |||
| D64053.1 | BAA10930.1 | |||
| U42361.1 | AAD09447.1 | |||
| U77916.1 | AAB54006.1 | |||
| U77917.1 | AAB54007.1 | |||
| X82635.1 | CAA57957.1 | |||
| Z79693.1 | CAB01957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32743 | Chr.12:70481471 - 70719432 on Build GRCh38 | Gain |
|
| nsv509472 | Chr.12:70606856 - 70653543 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map