Assay Details
Target Gene Details
Entrez Gene ID: | 9194 |
Gene Name: | solute carrier family 16 member 7 |
Gene Aliases: |
MCT2 |
Location: |
Chr.12:59595934-59789855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC16A7 | NM_001270622.1 | NP_001257551.1 | ||
NM_001270623.1 | NP_001257552.1 | |||
XM_011538989.2 | XP_011537291.1 | |||
AF049608.1 | AAC70919.1 | |||
DR000391.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1048949 | Chr.12:59580258 - 59647696 on Build GRCh38 | Gain |
![]() |
nsv1038100 | Chr.12:59589610 - 59624964 on Build GRCh38 | Loss |
![]() |
esv3629661 | Chr.12:59555455 - 59609269 on Build GRCh38 | Gain |
![]() |
esv3629653 | Chr.12:59417980 - 59610460 on Build GRCh38 | Gain |
![]() |
esv2762982 | Chr.12:59266090 - 59606354 on Build GRCh38 | Gain |
![]() |
nsv559050 | Chr.12:59087714 - 61442753 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
