Assay Details
Target Gene Details
| Entrez Gene ID: | 775 | 
| Gene Name: | calcium voltage-gated channel subunit alpha1 C | 
| Gene Aliases: | CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS | 
| Location: | Chr.12:1969677-2697949 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 5 | 
Target Gene Details
| Entrez Gene ID: | 100874234 | 
| Gene Name: | CACNA1C antisense RNA 4 | 
| Gene Aliases: | - | 
| Location: | Chr.12:2220537-2223481 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CACNA1C-AS4 | NR_046578.1 | |||
| BI830379.1 | ||||
| DB094045.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1040370 | Chr.12:1912229 - 2561851 on Build GRCh38 | Gain |  CACNA1C-IT3  CACNA1C-IT2  CACNA1C  CACNA1C-AS4  LINC00940  CACNA2D4  DCP1B | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top