Assay Details
Target Gene Details
Entrez Gene ID: | 105370070 |
Gene Name: | cervical cancer-associated transcript 8 |
Gene Aliases: |
- |
Location: |
Chr.12:128023788-128027166 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRAT8 | NR_131985.1 | |||
| DA425353.1 | ||||
| DA428755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3631127 | Chr.12:127897320 - 128034186 on Build GRCh38 | Gain |
|
| dgv1573n100 | Chr.12:126502891 - 128427644 on Build GRCh38 | Gain |
|

Genomic Map