Assay Details
Target Gene Details
Entrez Gene ID: | 64426 |
Gene Name: | SDS3 homolog, SIN3A corepressor complex component |
Gene Aliases: |
SAP45, SDS3 |
Location: |
Chr.12:118373189-118418035 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUDS3 | XM_011538671.2 | XP_011536973.1 |
More Information
Set Membership: |
|

Genomic Map