Assay Details
Target Gene Details
Entrez Gene ID: | 84305 |
Gene Name: | PYM homolog 1, exon junction complex associated factor |
Gene Aliases: |
PYM, WIBG |
Location: |
Chr.12:55901413-55927913 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYM1 | NM_001143853.1 | NP_001137325.1 | ||
| NM_032345.2 | NP_115721.1 | |||
| AJ459406.1 | CAD30677.1 | |||
| AK096922.1 | BAC04897.1 | |||
| BC006135.1 | AAH06135.1 | |||
| BC009627.1 | AAH09627.1 | |||
| BC014976.1 | AAH14976.1 | |||
| DB466388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832424 | Chr.12:55881442 - 56073133 on Build GRCh38 | Loss |
|
| nsv1051961 | Chr.12:55868436 - 56235643 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map