Assay Details
Target Gene Details
Entrez Gene ID: | 100129361 |
Gene Name: | small integral membrane protein 10 like 1 |
Gene Aliases: |
- |
Location: |
Chr.12:11171181-11176020 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMIM10L1 | NM_001271592.1 | 1 | 3327 | NP_001258521.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050050 | Chr.12:11039447 - 11283425 on Build GRCh38 | Loss |
|
| dgv1378n100 | Chr.12:11068113 - 11175532 on Build GRCh38 | Loss |
|
| esv2759879 | Chr.12:10817021 - 11561987 on Build GRCh38 | Loss |
|
| nsv1038277 | Chr.12:10908379 - 11351157 on Build GRCh38 | Loss |
|
| nsv428272 | Chr.12:10917246 - 11561987 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs375174709] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map