Assay Details
Target Gene Details
Entrez Gene ID: | 5800 |
Gene Name: | protein tyrosine phosphatase, receptor type O |
Gene Aliases: |
GLEPP1, NPHS6, PTP-OC, PTP-U2, PTPROT, PTPU2, R-PTP-O |
Location: |
Chr.12:15322257-15598331 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRO | NM_002848.3 | NP_002839.1 | ||
| NM_030667.2 | NP_109592.1 | |||
| XM_017019725.1 | XP_016875214.1 | |||
| AB209729.1 | BAD92966.1 | |||
| AK289784.1 | ||||
| BC035960.1 | AAH35960.1 | |||
| BC126201.1 | ||||
| BC126203.1 | ||||
| DA165109.1 | ||||
| DR000351.1 | ||||
| U20489.1 | AAA82892.1 | |||
| Z48541.1 | CAA88425.1 |
More Information
Set Membership: |
|

Genomic Map