Assay Details
Target Gene Details
Entrez Gene ID: | 5153 |
Gene Name: | phosphodiesterase 1B |
Gene Aliases: |
HEL-S-79p, PDE1B1, PDES1B |
Location: |
Chr.12:54549393-54579239 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDE1B | NM_000924.3 | NP_000915.1 | ||
NM_001288768.1 | NP_001275697.1 | |||
NM_001288769.1 | NP_001275698.1 | |||
XM_011538456.1 | XP_011536758.1 | |||
AB209200.1 | BAD92437.1 | |||
AK296422.1 | ||||
AK302899.1 | ||||
BC032226.1 | AAH32226.1 | |||
DA254686.1 | ||||
EU668337.1 | ||||
U56976.1 | AAC50769.1 | |||
U86078.1 | AAC51872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832421 | Chr.12:54522913 - 54691618 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
