Assay Details
Target Gene Details
Entrez Gene ID: | 102216268 |
Gene Name: | chondrogenesis-associated transcript |
Gene Aliases: |
CISTR-ACT, CISTRACT, re52431 |
Location: |
Chr.12:53750447-53757034 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CISTR | NR_104332.1 | |||
| HY042141.1 |

Genomic Map