Assay Details
Target Gene Details
Entrez Gene ID: | 51071 |
Gene Name: | deoxyribose-phosphate aldolase |
Gene Aliases: |
CGI-26, DEOC |
Location: |
Chr.12:15911172-16037381 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DERA | NM_001300779.1 | NP_001287708.1 | ||
| NM_015954.3 | NP_057038.2 | |||
| AF132960.1 | AAD27735.1 | |||
| AK024614.1 | ||||
| AK222541.1 | BAD96261.1 | |||
| AK296697.1 | ||||
| BC056234.1 | AAH56234.1 | |||
| BU928832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036638 | Chr.12:15912446 - 16358900 on Build GRCh38 | Loss |
|
| esv3892147 | Chr.12:15925316 - 16035459 on Build GRCh38 | Loss |
|
| nsv557636 | Chr.12:15927715 - 16010449 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map