Assay Details
Target Gene Details
Entrez Gene ID: | 81539 |
Gene Name: | solute carrier family 38 member 1 |
Gene Aliases: |
ATA1, NAT2, SAT1, SNAT1 |
Location: |
Chr.12:46183055-46269425 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC38A1 | NM_001077484.1 | NP_001070952.1 | ||
| NM_001278387.1 | NP_001265316.1 | |||
| NM_001278388.1 | NP_001265317.1 | |||
| NM_001278389.1 | NP_001265318.1 | |||
| NM_001278390.1 | NP_001265319.1 | |||
| NM_030674.3 | NP_109599.3 | |||
| XM_011538784.2 | XP_011537086.1 | |||
| XM_011538786.2 | XP_011537088.1 | |||
| XM_011538787.2 | XP_011537089.1 | |||
| XM_017019990.1 | XP_016875479.1 | |||
| AF247166.1 | AAG44546.1 | |||
| AF271070.1 | AAG39354.1 | |||
| AK027825.1 | BAB55394.1 | |||
| AK074758.1 | BAC11186.1 | |||
| AK074797.1 | BAC11215.1 | |||
| AK074851.1 | ||||
| AK074949.1 | BAC11310.1 | |||
| AK130314.1 | 1 | 2473 | ||
| AK225639.1 | ||||
| AK225778.1 | ||||
| AK308129.1 | ||||
| AK309410.1 | ||||
| AK313731.1 | ||||
| BC010620.1 | AAH10620.1 | |||
| BX647547.1 | ||||
| BX648495.1 | ||||
| BX648676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826360 | Chr.12:46121591 - 46204460 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map