Assay Details
Target Gene Details
Entrez Gene ID: | 7009 |
Gene Name: | transmembrane BAX inhibitor motif containing 6 |
Gene Aliases: |
BAXI1, BI-1, TEGT |
Location: |
Chr.12:49741510-49764934 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMBIM6 | NM_001098576.1 | NP_001092046.1 | ||
| NM_003217.2 | NP_003208.2 | |||
| XM_005269126.3 | XP_005269183.1 | |||
| AF033095.1 | AAB87479.1 | |||
| AK124849.1 | ||||
| AK130050.1 | ||||
| AK225804.1 | ||||
| AK299587.1 | ||||
| AK304577.1 | ||||
| AK312238.1 | ||||
| BC000916.1 | AAH00916.1 | |||
| BC036203.1 | ||||
| DA476870.1 | ||||
| DC319835.1 | ||||
| DC349385.1 |
More Information
Set Membership: |
|

Genomic Map