Assay Details
Target Gene Details
Entrez Gene ID: | 5149 |
Gene Name: | phosphodiesterase 6H |
Gene Aliases: |
ACHM6, RCD3 |
Location: |
Chr.12:14973022-14982697 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE6H | NM_006205.2 | NP_006196.1 | ||
| D45399.1 | BAA08241.1 | |||
| EL946662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951333 | Chr.12:14951167 - 14983766 on Build GRCh38 | Duplication |
|
| dgv1394n100 | Chr.12:14938693 - 14980786 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map